Learn More
Hexanes, mixed isomers, ACS, 98.5+%, Thermo Scientific Chemicals
£24.91 - £114.00
Chemical Identifiers
CAS | 92112-69-1 |
---|---|
Molecular Formula | C6H14 |
Molecular Weight (g/mol) | 86.18 |
MDL Number | MFCD00009520 |
InChI Key | VLKZOEOYAKHREP-UHFFFAOYSA-N |
Synonym | n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes |
PubChem CID | 8058 |
ChEBI | CHEBI:29021 |
IUPAC Name | hexane |
SMILES | CCCCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11405000
|
Thermo Scientific Alfa Aesar
033321.AP |
500 mL | |||||||
11367648
|
Thermo Scientific Alfa Aesar
033321.K2 |
1 L |
£37.50
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11377648
|
Thermo Scientific Alfa Aesar
033321.K4 |
2.5 L |
£74.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11347648
|
Thermo Scientific Alfa Aesar
033321.M1 |
4 x 1 L |
£114.00
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11357648
|
Thermo Scientific Alfa Aesar
033321.K7 |
4 L | |||||||
Description
Hexane mixed isomers is used in the laboratory to extract oil and grease contaminants from water and soil for analysis. As a non-polar solvent, it is used in chromatography. It plays important role in the preparation of glues for shoes, leather products, and roofing. It is also used for reactions that involve very strong bases such as the preparation of Grignard reagents. Also involved in the preparation of organolithiums compounds namely butyl lithium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
92112-69-1 | |
86.18 | |
VLKZOEOYAKHREP-UHFFFAOYSA-N | |
8058 | |
hexane |
C6H14 | |
MFCD00009520 | |
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
CHEBI:29021 | |
CCCCCC |
Specifications
-95°C | |
Colorless | |
65°C to 70°C | |
Liquid, Mixture Of 5 Isomers | |
C6H14 | |
UN1208 | |
VLKZOEOYAKHREP-UHFFFAOYSA-N | |
hexane | |
8058 | |
86.18 | |
ACS |
92112-69-1 | |
0.67g/mL | |
<−23° (−9°F) | |
1.3795 | |
MFCD00009520 | |
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes | |
CCCCCC | |
86.18 | |
CHEBI:29021 | |
≥98.5% | |
Hexanes, Mixed isomers |
Safety and Handling
GHS H Statement
H225-H304-H361-H373-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.
GHS P Statement
P210-P260-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEXANES
missing translation for 'tsca' : TSCA
RUO – Research Use Only