missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, mixed isomers, HPLC Grade, 99+%, Thermo Scientific Chemicals

£59.09

Specifications

Chemical Name or Material Hexanes
Melting Point -95°C
CAS 92112-69-1, 110-54-3, 96-14-0, 107-83-5, 96-37-7, 71-43-2
Color Colorless
Appearance Colorless liquid
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15442427
View Documents
Thermo Scientific Alfa Aesar
045652.K2
1 L
£59.09
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Hexanes, mixed isomers are used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Hexanes
92112-69-1, 110-54-3, 96-14-0, 107-83-5, 96-37-7, 71-43-2
Colorless liquid
<-23°C (-9°F)
Liquid
>99%
MFCD02179311
Miscible with ether,chloroform,alcohol,and acetone. Immiscible with water.
HPLC
-95°C
Colorless
65°C to 70°C
240°C (464°F)
1.3795
C6H14
160 hPa (120mm Hg) at 20°C
86.18
n-Hexane: 65.0%; 3-Methylpentane: 25.0%; 2-Methylpentane: 5.0%; Methylcyclopentane: 4.999%; Benzene: 0.001%
Videos
Safety and Handling

Safety and Handling

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c

H225-H304-H315-H319-H335-H336-H361f-H372

missing translation for 'dotInformation' : Hazard Class: 3, Packing Group: II

missing translation for 'einecsNumber' : 295-570-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Hexanes, mixed isomers, HPLC Grade

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Hexane, branched andlinear

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.