Learn More
Hexanoic acid, 99%, Thermo Scientific Chemicals
£27.20 - £126.00
Chemical Identifiers
CAS | 142-62-1 |
---|---|
Molecular Formula | C6H12O2 |
Molecular Weight (g/mol) | 116.16 |
MDL Number | MFCD00004421 |
InChI Key | FUZZWVXGSFPDMH-UHFFFAOYSA-N |
Synonym | caproic acid, capronic acid, n-hexanoic acid, n-caproic acid, butylacetic acid, hexoic acid, pentylformic acid, 1-hexanoic acid, n-hexylic acid, n-hexoic acid |
PubChem CID | 8892 |
ChEBI | CHEBI:30776 |
IUPAC Name | hexanoic acid |
SMILES | CCCCCC(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10115543
|
Thermo Scientific Acros
120702500 |
250 g | Glass bottle |
£27.20
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10102250
|
Thermo Scientific Acros
120705000 |
500 g | Glass bottle |
£40.30
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10306470
|
Thermo Scientific Acros
120700010 |
1 kg | Glass bottle |
£59.60
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10626204
|
Thermo Scientific Acros
120700025 |
2.5 kg | Glass bottle |
£126.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
142-62-1 | |
116.16 | |
FUZZWVXGSFPDMH-UHFFFAOYSA-N | |
8892 | |
hexanoic acid |
C6H12O2 | |
MFCD00004421 | |
caproic acid, capronic acid, n-hexanoic acid, n-caproic acid, butylacetic acid, hexoic acid, pentylformic acid, 1-hexanoic acid, n-hexylic acid, n-hexoic acid | |
CHEBI:30776 | |
CCCCCC(=O)O |
Specifications
473 to 493mg KOH/g | |
-3.0°C | |
0.9200g/mL | |
104°C | |
98.5% min. (GC) | |
CH3(CH2)4CO2H | |
MFCD00004421 | |
15, 1761 | |
caproic acid, capronic acid, n-hexanoic acid, n-caproic acid, butylacetic acid, hexoic acid, pentylformic acid, 1-hexanoic acid, n-hexylic acid, n-hexoic acid | |
FUZZWVXGSFPDMH-UHFFFAOYSA-N | |
hexanoic acid | |
8892 | |
3.23 mPa.s (20°C) | |
99% | |
Hexanoic acid |
142-62-1 | |
Colorless to Yellow | |
202.0°C to 203.0°C | |
Authentic | |
C6H12O2 | |
1.4150 to 1.4180 | |
02, 321 | |
0.92 | |
Solubility in water: 1.1% (20°C). Other solubilities: soluble in alcohol and ether | |
CCCCCC(=O)O | |
116.16 | |
CHEBI:30776 | |
116.16 | |
Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 205-550-7
- Hexanoic acid
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 C
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only