missing translation for 'onlineSavingsMsg'
Learn More

Hexyl alcohol, 98%, pure, Thermo Scientific Chemicals

Product Code. 10192290
Click to view available options
Quantity:
100 mL
1 L
2.5 L
10 L
25 L
Packaging:
Glass bottle
Metal Drum
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-27-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
MDL Number MFCD00002982
InChI Key ZSIAUFGUXNUGDI-UHFFFAOYSA-N
Synonym 1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinol
PubChem CID 8103
ChEBI CHEBI:87393
IUPAC Name hexan-1-ol
SMILES CCCCCCO

Specifications

Acidity 0.02mg KOH/g max.
Melting Point -52.0°C
Density 0.8136g/mL
Boiling Point 156.0°C to 157.0°C
Flash Point 59°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)5OH
Refractive Index 1.4170 to 1.4190
Quantity 100 mL
Beilstein 01, 407
Merck Index 15, 4734
Specific Gravity 0.8136
Solubility Information Solubility in water: 6g/L (25°C). Other solubilities: miscible with alcohol and ether
Viscosity 5.3 mPa.s (20°C)
Formula Weight 102.18
Percent Purity 98%
Grade Pure
Physical Form Liquid
Chemical Name or Material Hexyl alcohol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'