Learn More
Hexyl alcohol, 99%, anhydrous, AcroSeal™, Thermo Scientific Chemicals
£26.00 - £129.00
Identificadores de productos químicos
CAS | 111-27-3 |
---|---|
Molecular Formula | C6H14O |
Molecular Weight (g/mol) | 102.18 |
InChI Key | ZSIAUFGUXNUGDI-UHFFFAOYSA-N |
Synonym | 1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinol |
PubChem CID | 8103 |
ChEBI | CHEBI:87393 |
IUPAC Name | hexan-1-ol |
SMILES | CCCCCCO |
Código de producto | Marca | Quantity | Packaging | Precio | Cantidad y disponibilidad | ||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Quantity | Packaging | Precio | Cantidad y disponibilidad | ||||
11984611
|
Thermo Scientific Acros
433861000 |
100 mL | AcroSeal™ Glass Bottle |
£26.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11974611
|
Thermo Scientific Acros
433860010 |
1 L | AcroSeal™ Glass Bottle |
£129.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identificadores de productos químicos
111-27-3 | |
102.18 | |
1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinol | |
CHEBI:87393 | |
CCCCCCO |
C6H14O | |
ZSIAUFGUXNUGDI-UHFFFAOYSA-N | |
8103 | |
hexan-1-ol |
Especificaciones
111-27-3 | |
Colorless | |
156°C to 157°C | |
Authentic | |
C6H14O | |
1.4170 to 1.419 | |
15,4734 | |
1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinol | |
ZSIAUFGUXNUGDI-UHFFFAOYSA-N | |
hexan-1-ol | |
8103 | |
5.3 mPa.s (20°C) | |
99% | |
Hexyl alcohol |
-52°C | |
0.8136g/mL | |
59°C | |
98.5% min. (GC) | |
CH3(CH2)5OH | |
0.0005 % max. | |
0.8136 | |
Solubility in water: 6g/L (25°C). Other solubilities: miscible with alcohol and ether | |
CCCCCCO | |
102.18 | |
CHEBI:87393 | |
102.18 | |
Liquid |
Seguridad y manipulación
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously wit
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 203-852-3
- Hexyl alcohol
Palabra de advertencia
- Warning
Clasificación
- Flammable liquid Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Indicaciones de peligro
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H226-Flammable liquid and vapour.
Consejos de prudencia
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only