missing translation for 'onlineSavingsMsg'
Learn More

Hexylamine, 99%

Product Code. 10318440 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
100 mL
500 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
100mL
500mL
5mL
This item is not returnable. View return policy

Product Code. 10318440

Brand: Thermo Scientific Acros 204725000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-26-2 | C6H15N | 101.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-26-2
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.19
MDL Number MFCD00008240
InChI Key BMVXCPBXGZKUPN-UHFFFAOYSA-N
Synonym hexylamine, 1-aminohexane, n-hexylamine, 1-hexanamine, 1-hexylamine, mono-n-hexylamine, hexanamine, hexyl amine, unii-ci4e002zv8, hexyl-amine
PubChem CID 8102
ChEBI CHEBI:5712
IUPAC Name hexan-1-amine
SMILES CCCCCCN

Specifications

Melting Point -19.0°C
Color Colorless to Yellow
Density 0.7500g/mL
Boiling Point 130.0°C
Flash Point 27°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)5NH2
Refractive Index 1.4170 to 1.4190
Quantity 500 mL
Beilstein 04, 188
Fieser 07,170
Specific Gravity 0.75
Solubility Information Solubility in water: 14g/L (20°C).
Viscosity 0.83 mPa.s (20°C)
Formula Weight 101.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Hexylamine, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hexylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.