Learn More
Cole-Parmer™ HF Acid Eater and Spill Kit
Shop All Cole-Parmer Products
Click to view available options
Capacity (Metric):
18.927 L
Description
- Neutralizes hydrofluoric acid into safe organic salts
- Unlike caustic soda and soda ash, which forms poisonous bi-fluoride, HF Acid Eater completely destroys the hydrofluoric acid polymer
- This neutralizer has a 1:1 neutralization ratio as apposed to the 12:1 ratio of caustic soda.
- pH indicator additive eliminates the need for additional pH strips for followup testing
- Nonhazardous and noncorrosive, and is excellent for a variety of uses and applications
- Apply on horizontal, vertical, and perforated surfaces
- Also neutralizes vapors and reduces odors
Specifications
Specifications
| Capacity (English) | 5 gal. |
| Capacity (Metric) | 18.927 L |
| Type | Hydrofluric Acid Spill Kit |