missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HHLA3 Monoclonal Antibody (1F6)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524445
This item is not returnable.
View return policy
Description
Peptide Sequence: MFGACYKQPL KPSGSEPPAE ECRMTPRHAG CDVTEMQRIL SQPTFTEHLL RAV.
HHLA3 (HERV-H LTR-Associating 3) is an RNA Gene, and is affiliated with the lncRNA class. [GeneCards].
Specifications
| HHLA3 | |
| Monoclonal | |
| Unconjugated | |
| HHLA3 | |
| HERV-H LTR-associating 3; HERV-H LTR-associating protein 3; HHLA3; human endogenous retrovirus-H long terminal repeat-associating 3; human endogenous retrovirus-H long terminal repeat-associating protein 3 | |
| Mouse | |
| Protein A | |
| RUO | |
| 11147 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1F6 | |
| PBS with no preservative; pH 7.4 | |
| Q9XRX5 | |
| HHLA3 | |
| HHLA3 (NP_009002, 1 a.a. ~ 53 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction