missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Hi-Di™ Formamide

Product Code. 15803570 Shop All Applied Biosystems Products
Click to view available options
Quantity:
25 mL
4 x 5 mL
Unit Size:
25mL
Each
This item is not returnable. View return policy

Product Code. 15803570

Brand: Applied Biosystems™ 4311320

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly deionized (Hi-Di) formamide

Highly deionized (Hi-Di) formamide is used to resuspend samples before electrokinetic injection in capillary electrophoresis systems.

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -15°C to -25°C. Avoid multiple freeze-thaw cycles.
For Use With (Application) Sequencing
Product Line High-Di
Product Type Formamide
Quantity 25 mL
Reagent Type Formamide
Shipping Condition Dry Ice
Form Liquid
compliance-icons
Product Identifier
  • Hi-Di Formamide
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H360-May damage fertility or the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P314-Get medical advice/attention if you feel unwell.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : formamide

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.