Learn More
Applied Biosystems™ Hi-Di™ Formamide
Highly deionized (Hi-Di) formamide
Brand: Applied Biosystems™ 4440753
49.33 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.
67.69 GBP valid until 2025-12-31
Use promo code "24365" to get your promotional price.
Description
- Highly deionized (Hi-Di) formamide is used to resuspend samples before electrokinetic injection in capillary electrophoresis systems.
- The smaller configuration (5mL) minimizes the number of freeze/thaw cycles compared to the larger 25mL bottle
Order Info
Each

Specifications
High-Di™ | |
Formamide, 4x 5mL, Liquid, Formamide Reagent Type | |
4x 5 mL |
Store at -15 to -25°C. | |
Formamide |
Safety and Handling
- Formamide
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H351-Suspected of causing cancer.
- H360D-May damage the unborn child.
- H373-May cause damage to organs.
Precautionary Statement
- P202-Do not handle until all safety precautions have been read and understood.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : formamide
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.