Learn More
Alexandra™ Hi-Vis Waistcoat Jacket, Blue
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Description
- Double band and brace reflective tape
- Velcro™ fastening
- Conforms to EN471 class 2
- Up to 40° wash
- 100% polyester
Specifications
Specifications
Size | X-Large |
Gender | Unisex |
Color | Blue |
Style | SW22 |
Product Content Correction
Product Title
Alexandra™ Hi-Vis Waistcoat Jacket, Blue >
Spot an opportunity for improvement?Share a Content Correction