missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HIC (Hydrophobic Interaction Chromatography) Media
Features high chemical stability, properties to increase productivity and epoxy coupling
£134.00 - £135.00
Specifications
Flow Rate | 600 cm/Hoμr |
---|---|
For Use With (Application) | For Used in The Capture and Intermediate Stages of Protein PurifiCatIon |
Quantity | 25 mL |
Media Type | Highly Cross-Linked Agarose |
Max. Pressure | 300 kPa |
Product Code | Brand | Product Type | Ligand Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Ligand Type | Price | Quantity & Availability | ||||
11344063
|
Cytiva
17-5451-01 |
Capto Phenyl (High Sub) media | Phenyl |
£135.00
25mL |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
11638873
|
Cytiva
17-5459-01 |
Capto Butyl media | Butyl |
£134.00
25mL |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
Description
- High flow rate, high capacity and low backpressure reduce cycle times
- Chemically stable to commonly used aqueous buffers
- Epoxy chemical coupling
- Capto Phenyl (high sub) has a hydrophobic ligand of phenyl and is a hydrophobic interaction chromatography medium used for protein purification
- Capto Butyl has a hydrophobic ligand of butly and a highly cross-linked agarose matrix
Used in capture and intermediate stages of protein purification
Specifications
600 cm/Hoμr | |
25 mL | |
300 kPa | |
3 to 13 | |
20% Ethanol | |
20 cm |
For Used in The Capture and Intermediate Stages of Protein PurifiCatIon | |
Highly Cross-Linked Agarose | |
75 μm | |
4°C to 30°C | |
27 mg BSA/mL |