Learn More
Applied Biosystems™ High-Capacity RNA-to-cDNA™ Kit
The High Capacity RNA-to-cDNA Kit is a streamlined reverse transcription kit designed for optimum performance with TaqMan Gene Expression Master Mix, Power SYBR Green PCR Master Mix, and other PCR enzymes.
Brand: Applied Biosystems™ 4388950
1639.65 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 0.02200kg
Includes: 1 x 500 uL enzyme mix (MuLV and RNase inhibitor protein) (20X), 5 x 1 mL RT Buffer Mix (2X) (includes dNTPs, random octamers, and oligo dT-16)
Description
The High Capacity RNA-to-cDNA Kit is a streamlined reverse transcription kit designed for optimum performance with TaqMan Gene Expression Master Mix, Power SYBR Green PCR Master Mix, and other PCR enzymes. The components of the two-tube kit work together to provide sensitive and specific reverse transcription across a broad range of template amounts.
Features of the High Capacity RNA-to-cDNA Kit include:
• Fast reaction time (typically 30–60 min)
• Convenient and simple workflow with few pipetting steps
• Reliable reverse transcription of both abundant and limited targets
• Optimized two-step protocol enables multiple PCR reactions from a single RT reaction
Streamline cDNA synthesis while maintaining detection sensitivity
Amplification of a dilution series of the RNA concentration reference standard demonstrates exceptional reverse transcription and PCR efficiency across 11 orders of magnitude. This wide dynamic range allows one set of reaction conditions to detect transcripts from highly active as well as weakly expressed genes. The two-tube formulation reduces reaction time and requires fewer pipetting steps than with the High Capacity cDNA Reverse Transcription Kit, while maintaining high performance.
Specifications
• 20X Enzyme mix (1 x 500 μL) |
|
30 to 60 min. | |
37°C | |
Yes | |
Real Time PCR (qPCR), RT-PCR | |
500 Reactions | |
Reverse Transcription | |
RNA |
Kit | |
Reverse Transcription | |
MultiScribe | |
Dry Ice | |
First-Strand cDNA | |
Premixed Components | |
Up to 7 kb |
For Research Use Only. Not for use in diagnostic procedures.