missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ High Capacity Section Dryer
Provide maximum drying capacity for high workload laboratories with the Epredia™ High Capacity Section Dryer.
£3595.00
Specifications
Max. Temperature (Metric) | 85°C |
---|---|
Min. Temperature (Metric) | Room Temp: 85°C (±1°C) at 25°C ambient |
Description
Features
- Small footprint
- Anti-microbial paint finish (silver ion) to inhibit growth of bacteria
- Miniature circuit breakers for safety
Heating System
- Fan-assisted heating reduces section drying time
- Ultra-fast heating system for rapid warm up
- Digital temperature control guarantees heat accuracy within the chamber
Specifications
85°C | |
Room Temp: 85°C (±1°C) at 25°C ambient |