missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ High-Density Polyethylene Atomizers

Catalog Number 12005302
40.39 GBP valid until 2024-12-31
Use promo code "23264" to get your promotional price.


Click to view available options
Capacity (Metric):
400 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Vary delivery by turning nozzle from a fine spray to a solid stream.

  • HDPE construction
  • White
  • Turn nozzle to vary delivery from a fine spray to a solid stream
Capacity (Metric) 400 mL
Color White
For Use With (Application) Atomising
Material HDPE
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'