missing translation for 'onlineSavingsMsg'
Learn More
Learn More
High Five™ Microporous Lab Coats
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Description
Breathability with durability
- Cloth-like, microporous material
- General purpose protection from dirt, grime, splashes and spills
- Economical and lightweight
- Low linting and antistatic
- Elastic wrists, five-snap closure and two pockets
- White
Specifications
Specifications
Product Type | General Purpose Lab Coat |
Size | Large |
Gender | Unisex |
Material | Microporous Film |
Color | White |
Closure Type | Snaps |
No. of Pockets | 2 |
For Use With (Application) | General purpose protection from dirt, grime, splashes, and spills |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'