missing translation for 'onlineSavingsMsg'
Learn More

Chemgene™ High Level Surface Disinfectant

Product Code. p-8010821
Click to view available options
Volume (Metric):
5 L
Unit Size:
5 L
This item is not returnable. View return policy

Product Code. 15176686

Brand: Chemgene™ XTM308

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Combines enhanced active ingredient molecules with micelle cleaning technology to provide swift penetration of the cell walls to ensure rapid cell death. Chemgene™ High Level Surface Disinfectant ensures that cell death is achieved rather than just suspending activity.

  • Biodegradable
  • Non-corrosive
  • Long shelf life
  • Diluted concentrate lasts up to 6 months
  • Compliant with REACH regulations and the Biocidal Product directive

Specifications

Appearance Blue
Quantity 1 Ea.
Volume (Metric) 5 L
compliance-icons
Product Identifier
  • CHEMGENE HLD4L CONCENTRATE
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P315-Get immediate medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : alcohol C9-C11, bis(3-aminopropyl) dodecylamine, didecyldimethyl ammonium chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.