Learn More
UVP High-Performance Uv Variable Intensity Transilluminator
Brand:  UVP 95-0422-02
Code : 04
Additional Details : Weight : 7.35000kg
Product Code. 12873928
  • £1565.00 / Each
Estimated Shipment: 27-09-2024
to see stock.
Add to basket
Description
Description
UVP's high performance transilluminators feature 25W tubes and a specially constructed filter that provides the most uniform light emission possible from a tube based unit.
The units have a stainless steel top that is easy to clean and a hinged UV blocking lid to protect the user. High, medium and low intensity settings make the unit suitable for both visualisation and band cutting. An internal fan improves the tube lifetime.Dimensions: 337mm x 486mm x 143mm