missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan™ High Pressure Syringe
Designed for high pressure gas sampling
Brand: Trajan™ 008171
This item is not returnable.
View return policy
Description
- Designed for high pressure gas sampling (rated to 500psi)
- Fitted with a gas tight ON/OFF valve and high pressure 7cm, 23 gauge needle
Specifications
23 G | |
0.63 mm | |
High Pressure Syringe | |
1 mL |
0.37 mm | |
70 mm | |
1/Ea. | |
Sleeved Side Hole |