Learn More
Idex High-Pressure, Zero Dead Volume Unions
PEEK ZDV unions provide accurate results
Brand:  Idex 02014-37
 View more versions of this product
Product Code. 13010410
  • £106.00 / Each
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Inert PEEK fluid path
  • 10-32 coned configuration
Specifications
Specifications
Union
6000 psi
PEEK
0.01 in.
0.071 μL
1/16 in. OD Tubing