Learn More
Greiner Bio-One High Profile Polystyrene Microplate Lids
For use with non TC-treated 96-well microplates. Greiner Bio-one High Profile Polystyrene Microplate Lids is free of detectable DNase, RNase, human DNA.
Brand: Greiner Bio-One 656171
Additional Details : Weight : 1.00000kg
Description
- Free of detectable DNase, RNase, human DNA
- Non-Pyrogenic, non-cytotoxic
- Offer protection against contamination and evaporation during sample storage and cell cultivation
Specifications
Clear | |
Sterile | |
Yes | |
1 |
Polystyrene | |
9 mm | |
100 |
For Research Use Only. Not for use in diagnostic procedures, unless otherwise indicated.