missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva High Purity rNTP Solutions
Use for in vitro transcription or for reactions requiring rNTPs of high purity. Cytiva Lifescience™ High Purity rNTP Solutions provide easy-to-use, time-saving solutions.
£75.04
Specifications
Format | Liquid |
---|---|
Description | Easy-to-use, time-saving solutions for in vitro transcription or for reactions requiring rNTPs of high purity. |
pH | 7.5 |
Concentration | 100 mM |
Purity | ≥98% triphosphate |
Product Code | Brand | Format | Product Type | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Format | Product Type | Quantity | Price | Quantity & Availability | |||
10136184
|
Cytiva
27-2066-01 |
Liquid | CTP, 100 mM solution | 25 μmol |
£75.04
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10784737
|
Cytiva
27-2076-01 |
Liquid | GTP, 100 mM solution | 25 μmol |
£75.04
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10004164
|
Cytiva
27-2086-01 |
Liquid | UTP, 100 mM solution | 25 μmol |
£75.04
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10676505
|
Cytiva
27-2056-01 |
Liquid | ATP, 100 mM solution | 25 μmol |
£75.04
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Each lot is tested for ribonuclease contamination to ensure success in your application
- High purity: >98% triphosphate
Specifications
Liquid | |
7.5 | |
≥98% triphosphate | |
Colorless |
Easy-to-use, time-saving solutions for in vitro transcription or for reactions requiring rNTPs of high purity. | |
100 mM | |
25 μmol | |
in-vitro transcription, amplification, dideoxy sequencing, labeling, mutagenesis, cDNA synthesis and expression profiling |