missing translation for 'onlineSavingsMsg'
Learn More

Cytiva High Purity rNTPs: Solid

Easy-to-use, time-saving solutions for in vitro transcription or for reactions requiring rNTPs of high purity

Brand:  Cytiva 27-2000-04

 View more versions of this product

Product Code. 10419964

  • £533.00 / 1g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Each lot is tested for ribonuclease contamination to ensure success in your application
  • High purity: >98% triphosphate

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Solid
7.5
≥98% triphosphate
White
Easy-to-use, time-saving solutions for in vitro transcription or for reactions requiring rNTPs of high purity.
GTP, sodium, crystalline
1 g
in-vitro transcription, amplification, dideoxy sequencing, labeling, mutagenesis, cDNA synthesis and expression profiling
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers