missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva High Purity rNTPs: Solid
Easy-to-use, time-saving solutions for in vitro transcription or for reactions requiring rNTPs of high purity
Brand: Cytiva 27-1006-01
This item is not returnable.
View return policy
Description
- Each lot is tested for ribonuclease contamination to ensure success in your application
- High purity: >98% triphosphate
Specifications
Solid | |
551.15 | |
≥98% | |
White |
7.5 | |
ATP | |
5 g | |
in-vitro transcription, amplification, dideoxy sequencing, labeling, mutagenesis, cDNA synthesis and expression profiling |