Learn More
Chemetrics™ High Range COD Photometer
Brand:  Chemetrics™ A7325
Product Code. 11752103
  • £700.00 / Each
Explore more special offers
Specifications
CHEMetrics 0-1500 ppm HR COD vials (Mfr. Nos. K-7360S, K-7365, K-7361S, K-7366) and 0-15,000 ppm HR+ COD vials (Mfr. Nos. K-7370S, K-7375, K-7371S, K-7376)
4 AAA batteries
COD Photometer (high range)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products