Learn More
Thermo Scientific™ High Select™ Depletion Spin Columns
Decrease albumin, immunoglobulins, and other high-abundance proteins from human serum or plasma in preparation for mass spectrometry analysis, or 1D and 2D gel electrophoresis using High Select depletion spin columns or resin.
Brand: Thermo Scientific™ A36366
Additional Details : Weight : 0.90720kg
Description
Analysis of human fluids is often complicated by the presence of high concentrations of serum proteins, including albumin and IgG that can make up more than 70% of total serum protein. Removal of albumin, immunoglobulins, and other abundant proteins is essential for the study of low-abundance proteins. Thermo Scientific High Select HSA/Immunoglobulin and Top 14 Abundant Protein depletion spin columns and resins enable depletion of the most abundant proteins in human serum and plasma for improved detection of low-abundance proteins using mass spectrometry analysis or 1D or 2D gel electrophoresis.
• Flexible—resin provided in mini and midi column formats as well as bulk for custom filling options• Optimized—removes >95% of IgG and >95% of albumin (HSA/Immunoglobulin); removes >95% of IgG and >95% of albumin, plus 12 other abundant proteins (Top 14)
• Fast—process samples in ∼10 minutes
• Economical—cost-effective spin columns are priced for single use
• Reduced carry-over—one-time use prevents protein carryover and experimental variability
High Select HSA/Immunoglobulin Depletion Resin uses highly specific, immobilized anti-HSA and anti-immuoglobulin antibodies (IgG, IgA, IgM, IgD, and IgE) to remove human serum albumin (HSA) and all major subclasses of immunoglobulins from serum and plasma. High Select Top14 Abundant Protein Depletion Resin uses highly specific immobilized antibodies to remove human serum albumin (HSA), albumin, IgG, IgA, IgM, IgD, IgE, kappa and lambda light chains, alpha-1-acidglycoprotein, alpha-1-antitrypsin, alpha-2-macroglobulin, apolipoprotein A1, fibrinogen, haptoglobin, and transferrin from serum, plasma, or spinal fluids. The resins are provided in an economical and convenient spin column format specifically designed for one-step processing and single use or bulk format for custom filling.
Specifications
HSA/Immunoglobulin Depletion Mini Spin Columns | |
Column | |
Store at 2–8°C. | |
Wet Ice | |
Protein | |
Protein Depletion |
Mass Spectrometer | |
High Select™ | |
Unlabeled | |
Plasma, Serum | |
Mini Spin Column | |
24 Columns |