missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wheaton™ High Temperature Autoclave Bag
Autoclave bag ideal for safe biological waste disposal.
Brand: Wheaton™ 370110
71.24 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Can be used at high temperatures
- For inactivation of the most resistant biological samples

Specifications
Biohazard Autoclave Bag | |
Single-use | |
Autoclave | |
Non-sterile | |
200 | |
415 x 600 mm |
Autoclavable | |
Autoclaving lab waste | |
Polypropylene | |
Biohazard | |
Up to 135°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Wheaton™ High Temperature Autoclave Bag > Dimensions (L x W): 415 x 600 mm
Spot an opportunity for improvement?Share a Content Correction