missing translation for 'onlineSavingsMsg'
Learn More

Nabertherm™ High-Temperature Furnace, MoSi2 heating

Designed as tabletop models, these compact high-temperature furnaces have a variety of advantages. The first-class workmanship using high-quality materials, combined with ease of operation, make these furnaces all-rounders in research and the laboratory.

Brand:  Nabertherm™ LHT216PN

Code : NEW

Additional Details : Weight : 88.00000kg

 View more versions of this product

Product Code. 15581082

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Controller: P470
Specifications

Specifications

P470
2 L
90 x 150 x 150 mm
Lift door
1600°C
470 x 630 x 760+260 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers