Learn More
Nabertherm™ High-Temperature Furnaces with Scale for Determination of Combustion Loss and Thermogravimetric Analysis (TGA)
These high-temperature furnaces are specially developed to determine combustion loss during annealing and for thermogravimetric analysis in the lab. The complete system consists of the high-temperature furnace, a table frame, precision scale with feedthroughs into the furnace and powerful software.
Brand:  Nabertherm™ 001711560
Additional Details : Weight : 120.00000kg
 View more versions of this product
Product Code. 17397829
  • £17037.65 / Each
Estimated Shipment: 01-10-2024
to see stock.
Add to basket
Description
Description
  • High-quality molybdenum disilicide heating elements
  • Dual shell housing made of textured stainless steel sheets with additional fan cooling for low surface temperature
  • Adjustable air inlet
  • Exhaust air opening in the roof
  • 4 scales available for different maximum weights and scaling ranges
  • Process control and documentation for temperature and combustion loss via VCD software package for monitoring
Specifications
Specifications
P570
Up to 1600°C
370 x 655 x 890 mm
5000 W
16 L
150 x 150 x 150 mm
400 V