Learn More
High Vacuum Suction Machine Sam15 Mobile Unit
Brand:  N/A SAM15/01010101
Encompass
Product Code. 12947934
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
Inter-uterine aspirator with 1 x 2L autoclavable cannister 220-240V 50/60Hz 450mm x 200mm x 380mm (w x d x h) 30L/min flow rate, 0 to 650mmHg vacuum
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
High Vacuum Suction Machine Sam15 Mobile Unit