Learn More
Sonics Materials™ High Volume Continuous Flow Cell
Brand: Sonics Materials™ 630-0583
Code : NEW
Additional Details : Weight : 5.80000kg
Specifications
500 mL | |
Stainless Steel Housing, Titanium Alloy Ti-6Al-4V Probe |