missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiLoad™ 16/600 Superdex™ 30 pg Prepacked Column

Designed for high-resolution preparative gel filtration chromatography separating peptides

Brand:  Cytiva 28-9893-31


Product Code. 11350342

  • £2298.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket

Includes: Dismantling tool, support screen, net ring (10μm), O-ring, stop plugs, and instructions.
This item is not returnable. View return policy

Description

Description

  • Prepacked XK columns
  • 1/16 in. fittings for convenient connections to KTA design systems
  • High resolution with short run times and good recovery
  • High capacity at high flow rates
  • Separations can be scaled up to production levels
  • Prepacked for convenience and reproducibility
  • Produced by covalent bonding of dextran to highly cross-linked agarose
  • Separation properties of these media are determined by the dextran component
  • BioProcess medium combines high mechanical strength with high hydrophilicity, allowing high flow rates, and minimal nonspecific interactions

  • Maximum pressure: 42psi
  • Bed dimensions: 16 × 600mm
  • Flow rate: <1.0mL/min.
  • Bed volume: 120mL
  • Flow velocity: 10 to 50cm/hr.
  • Sample volume: 5mL
  • Theoretical plates: >13000m-1
  • Complete pack size: 1 × 120mL
Media
  • pH Stability working range: 3 to 12
  • pH Stability cleaning: 1 to 14
  • Average particle size: 34μm
  • Matrix: composite of cross-linked agarose and dextran
Column
  • Material (column tube): borosilicate glass
  • Porosity: 10μm
  • Bed support: net
  • Material (Bed support): polypropylene (PP) (reinforced glass fiber), polyamide (PA)
  • Material (Column hardware): borosilicate glass, polyamide, fluororubber, ethylene propylene
  • diene (EPDM), ETFE, superpolyoxymethylene
  • Outlet I.D.: 0.8mm (0.031 in.)
  • Column I.D.: 16mm (0.62 in.)
  • Inlet I.D.. 0.8mm (0.031 in.)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Superdex 30 PG size exclusion chromatography media prepacked into HiLoad 26/600 column
16 mm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiLoad 16/600 Superdex 30 pg

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 2

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-18 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers