missing translation for 'onlineSavingsMsg'
Learn More

Hippuric acid, 98%

Product Code. p-143237 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
2.5kg
500g
5g
This item is not returnable. View return policy

Product Code. 10541201

Brand: Thermo Scientific Acros 150270050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 495-69-2 | C9H9NO3 | 179.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 495-69-2
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.18
MDL Number MFCD00002692
InChI Key QIAFMBKCNZACKA-UHFFFAOYSA-N
Synonym hippuric acid, n-benzoylglycine, glycine, n-benzoyl, benzoylglycine, benzamidoacetic acid, benzoylaminoacetic acid, hippurate, hippursaeure, phenylcarbonylaminoacetic acid, benzamidoessigsaeure
PubChem CID 464
ChEBI CHEBI:18089
SMILES OC(=O)CNC(=O)C1=CC=CC=C1

Specifications

Melting Point 188.0°C to 191.0°C
Color White
Loss on Drying 1% max. (105°C)
Infrared Spectrum Authentic
Assay Percent Range 1% max. (Benzoic acid) (HPLC)
Packaging Glass Bottle
Linear Formula C6H5CONHCH2CO2H
Quantity 5 g
Beilstein 09,225
Fieser 01,432
Merck Index 15,4753
Solubility Information Solubility in water: 1g/250 mL cold water. Other solubilities: freely soluble in hot water,1g/100mL chloroform- 1g/400mL ether,1g/60mL amyl alcohol,freely soluble in hot ether,pract.insoluble in benzene,cs2 and petr.ether
IUPAC Name 2-benzamidoacetic acid
Formula Weight 179.17
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Hippuric acid, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.