Learn More
Cytiva HiPrep™ 26/10 Desalting Columns
Provide reliable and reproducible desalting and buffer exchange with sample sizes of up to 15mL per column. Cytiva HiPrep 26/10 Desalting Column is a convenient, easy-to-use column prepacked 53mL column with Sephadex G-25 Fine for desalting and buffer exchange.
Brand: Cytiva 17-5087-01
532.95 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : GE
Additional Details : Weight : 0.24300kg
Description
- Saves time and labor as replacement for dialysis
- Two to four columns can be used in series for sample sizes 30 to 60mL
- Compatible with a laboratory pump or a chromatography system
- Fittings are 1/16 in. for easy connection to AKTAdesign systems
- Long-term and working pH range 2 to 13
- Maximum flowrate: 15mL/min.
Specifications
HiPrep 26/10 Desalting are convenient, easy-to-use, 53 mL columns prepacked with Sephadex G-25 Fine resin for desalting and buffer exchange. | |
Laboratory pump or chromatography system such as AKTA design | |
1.02 in. | |
4°C to 30°C in 20% Ethanol | |
No |
HiPrep 26/10 Desalting | |
>20 μm (dry), 17 to 132 μm (wet) | |
53 mL | |
26 mm |