missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiPrep™ 26/10 Desalting Columns
Provide reliable and reproducible desalting and buffer exchange with sample sizes of up to 15mL per column. Cytiva HiPrep 26/10 Desalting Column is a convenient, easy-to-use column prepacked 53mL column with Sephadex G-25 Fine for desalting and buffer exchange.
Brand: Cytiva 17-5087-02
This item is not returnable.
View return policy
Description
- Saves time and labor as replacement for dialysis
- Two to four columns can be used in series for sample sizes 30 to 60mL
- Compatible with a laboratory pump or a chromatography system
- Fittings are 1/16 in. for easy connection to AKTAdesign systems
- Long-term and working pH range 2 to 13
- Maximum flowrate: 15mL/min.

Specifications
HiPrep 26/10 Desalting are convenient, easy-to-use, 53 mL columns prepacked with Sephadex G-25 Fine resin for desalting and buffer exchange. | |
Laboratory pump or chromatography system such as AKTA design | |
1.02 in. | |
4°C to 30°C in 20% Ethanol | |
Reusable |
HiPrep 26/10 Desalting | |
>20 μm (dry), 17 to 132 μm (wet) | |
53 mL | |
26 mm | |
4 x 53 mL |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HiPrep 26/10 Desalting, 4 pack
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16,5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva HiPrep™ 26/10 Desalting Columns > 4 x 53mL
Spot an opportunity for improvement?Share a Content Correction