missing translation for 'onlineSavingsMsg'
Learn More

Cytiva His Mag Sepharose™ excel Chromatography Media

His Mag Sepharose excel magnetic beads are designed for small-scale purification and screening of histidine-tagged proteins secreted in eukaryotic cell culture.

Brand:  Cytiva 17371220


 View more versions of this product

Product Code. 12691206

  • £173.00 / Pack of 2
Estimated Shipment: 26-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Samples that usually cause stripping of metal ions can therefore be added to the medium with retained protein-binding capacity and without pretreatment.

  • His Mag Sepharose excel is a magnetic IMAC medium precharged with nickel ions that are very strongly bound to a chelating ligand.
  • The medium enables an efficient and simplified workflow for capture, purification, and reproducible screening of histidine-tagged proteins.
  • Simplify small-scale purification and expression screening of histidine-tagged proteins secreted in eukaryotic cell culture.
  • Purify histidine-tagged proteins directly from samples that contain metal ion-stripping agents and save time and effort.
  • Increase target protein yield and decrease degradation through reduced and simplified sample handling.
  • Screen histidine-tagged proteins in parallel with high reproducibility and reliable results.
  • Capture histidine-tagged protein from different sample volumes; from low to high milliliter scale.
  • Simplify handling using visible and dense Sepharose-based magnetic beads.
  • Proteins are expressed and secreted into cell culture supernatants from eukaryotic cells such as insect cells or Chinese Hamster Ovary (CHO) cells.
  • The high density of the beads allows rapid capture by magnetic devices while the visibility of the beads ensures reliable collection of the bound histidine-tagged proteins during the purification procedure.
  • The base matrix is based on Sepharose medium with magnetite incorporated.
  • His Mag Sepharose is available as 10% medium slurry in three different pack sizes (2 x 1ml, 5 x 1ml, and 10 x 1ml) and can be used together with different test tubes and magnetic racks.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

2 to 12
4°C to 30°C
2 x 1 mL
Chromatography Media
Highly cross-linked spherical Agarose including Magnetite
63 μm
Designed to simplify small-scale purification and screening of His-tagged Proteins_x000D_ secreted in eukaryotic cell culture, used for functional and structural studies research in academia, institutes and discovery Pharma
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • EUH208-Contains nickel. May produce an allergic reaction.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P235-Keep cool.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : triiron tetraoxide,ethanol, nickel
  • UNKNOWN AQUATIC-Contains 50% of components with unknown hazards to the aquatic environment.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers