missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiScreen™ Blue, Ni and DEAE Sepharose™ Prepacked Fast Flow Columns
Used for method optimization and parameter screening
£104.00 - £171.00
Specifications
Bed Volume | 4.7mL |
---|---|
Diameter (English) Inner | 0.3 in. |
Diameter (Metric) Inner | 7.7mm |
Quantity | 1 x 4.7mL |
Height (English) | 3.93 in. |
Product Code | Brand | Buffer | Flow Rate | pH Range | Content And Storage | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Buffer | Flow Rate | pH Range | Content And Storage | Price | Quantity & Availability | ||
12305683
|
Cytiva
28-9782-44 |
0.1M HCl, 0.1M NaOH, 8M Urea | <450cm/hr. | 3 to 12 | 4° to 30°C |
£155.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
||
11603234
|
Cytiva
28-9782-43 |
70% Ethanol, 6M Guanidine Hydrochloric, 8M Urea | <600cm/hr. | 4 to 12 | 4° to 8°C |
£171.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
||
11370322
|
Cytiva
28-9782-45 |
1M NaOH, 8M Urea, 8M Guanidine Hydrochloride, 70% Ethanol | <450cm/hr. | 2 to 12 | 4° to 30°C |
£104.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
||
Description
- Based on 6% highly cross-linked agarose with an average bead size of 90μm
- Processed flow properties and high loading capacities
- Bed height of 10cm is designed to allow method optimization and parameter screening
- Easily connected in series to achieve 20cm bed height
- Small bed volume gives fast results and minimal sample/buffer consumption
- Combines high capacity with high flow velocity and low backpressure to reduce process cycle times and increase productivity
- Reproducible results, scalable to BioProcess™ columns packed with the same chromatography media using the same linear fluid velocity
- Bed volume: 4.7mL
- Bed dimension: 7.7 × 100mm
- Matrix: 6% cross-linked agarose beads
- Average particle size: 90μm
- Flow velocity: <600cm/hr or <450cm/hr
- Material (Column hardware and column tube): polypropylene
- Column I.D.: 7.7mm
Blue Sepharose
- Prepacked with BioProcess chromatography medium Blue Sepharose 6 Fast Flow
- Purification of albumin, enzymes (including NAD+ and NADP+), coagulation factors, interferons and related proteins
- Prepacked with Nickel Sepharose 6 Fast Flow
- Purification of histidine-tagged proteins with Immobilized Metal ion Affinity Chromatography (IMAC)
- Prepacked with DEAE Sepharose Fast Flow
- Weak anion exchanger (diethylaminoethyl group) BioProcess™ medium
Specifications
4.7mL | |
7.7mm | |
3.93 in. | |
Polypropylene | |
45 to 165μm |
0.3 in. | |
1 x 4.7mL | |
100mm | |
1.5 bar | |
6% Cross-linked Agarose |
Safety and Handling
Signal Word
Hazard Category
- Warning
Hazard Category
- Flammable liquid Category 3