missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiScreen™ Capto™ Adhere Impres Multimodal Chromatography Column
GREENER_CHOICE

Ready-to-use column for method optimization and parameter screening

Brand:  Cytiva 17371520

173.85 GBP valid until 2024-12-31
Use promo code "21805" to get your promotional price.



Product Code. 15171546

  • £183.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket

Includes: Certificate of Analysis
This item is not returnable. View return policy

Description

Description

  • Prepacked with different BioProcess™ media for convenient process development
  • Bed height: 10cm
  • Easily connected in series to achieve 20cm bed height
  • Small bed volume for fast results and minimal sample/buffer consumption
  • Reduces sample cost and buffer consumption
  • Binds proteins at high or low ionic strength
  • Efficient removal of aggregates, viruses, and main contaminants in MAb processes
  • High-resolution beads and ligand selectivity ensures high yield

  • Reproducible results
  • Allows effective, high-resolution polishing of MAb

Avoid using oxidizing agents, anionic detergents.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Certificate of analysis
Anion
40 μm
40 μm
HiScreen
MAbs Purification
43.51 psi
Chromatography Column
Polypropylene
7.7 mm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiScreen Capto adhere ImpRes

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.