missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiScreen™ Capto™ MMC ImpRes Multimodal Chromatography Column

Ready-to-use column for method optimization and parameter screening

Brand:  Cytiva 17371620


Product Code. 15101556

  • £169.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Prepacked with different BioProcess™ media for convenient process development
  • Bed height: 10cm
  • Easily connected in series to achieve 20cm bed height
  • Small bed volume for fast results and minimal sample/buffer consumption
  • Displays broad pH/salt operational window
  • Enables use of platform approach to MAb process development
  • Efficient removal of aggregates, viruses, and main contaminants in MAb processes
  • High-resolution beads and ligand selectivity ensures high yield

  • Reproducible results
  • Allows effective, high-resolution polishing of MAb
  • Binds proteins at high or low ionic strength
  • Reduces sample cost and buffer consumption

Avoid using oxidizing agents, cationic detergents.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

HPLC, Parameter Screening, Optimization Method
300,000 psi
40 μm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Prepacked column, HiScreen Capto MMC ImpRes 4.7mL

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers