missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiScreen™ Capto™ SP ImpRes Prepacked IEX Columns
Ideal for method optimization and parameter screening
Brand: Cytiva 17-5468-15
This item is not returnable.
View return policy
Description
- Prepacked with Capto SP ImpRes, a strong cation exchanger
- High-flow agarose matrix
- Small bed volume of 4.7mL gives fast results and minimal sample/buffer consumption
- Easily connected in series to achieve 20cm bed height
- Bed height of 10cm to allow method optimization and parameter screening

Specifications
Capto SP ImpRes |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HiScreen Capto SP ImpRes, 4.7 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva HiScreen™ Capto™ SP ImpRes Prepacked IEX Columns > Bed volume: 4.7mL; Bed dimension: 7.7 x 100mm
Spot an opportunity for improvement?Share a Content Correction