missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiScreen™ MabSelect™, Xtra™, SuRe™, SuRe LX™ Prepacked Columns

Designed for method optimization and parameter screening for capture of monoclonal antibodies. Cytiva HiScreen™ MabSelect™, Xtra™, SuRe™, SuRe LX™ Prepacked Columns come ready-to-use for convenience.

Brand:  Cytiva 17-5474-15


 View more versions of this product

Product Code. 11603084

  • £1046.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Ready-to-use columns for convenient process development
  • Rigid, highly cross-linked agarose matrix
  • Bed height of 10cm is designed to allow method optimization and parameter screening
  • Easily connected in series to achieve 20cm bed height
  • Small bed volume (4.7mL) gives fast result and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity

MabSelect

  • For capture of MAbs from large sample volumes
  • Prepacked with protein A-derived media
  • Compatible with high flow rates and high pressure when scaling up
MabSelect Xtra
  • Increased binding capacity enables purification of MAbs from high-level expression feedstock
  • Also optimized for Fc-fusion proteins
  • Very low, unspecific binding due to high ligand selectivity and matrix hydrophilicity
  • Prepacked with protein A-derived media
MabSelect SuRe (Superior Resistance) and MabSelect SuRe (Superior Resistance) LX
  • With novel, alkali-tolerant recombinant protein A ligand that provides greater stability and allows use of 0.1 to 0.5M sodium hydroxide for cleaning-in-place (CIP)
  • Enhanced protease resistance resulting in lower ligand leakage
  • High dynamic binding capacity

TRUSTED_SUSTAINABILITY
Specifications

Specifications

8M Urea, 6M Guanidine Hydrochloride, 2% Benzyl Alcohol or 20% Ethanol
Stoppers at the inlet and outlet
Filter
≈60mg Human IgG
HiScreen MabSelect SuRe LX Prepacked Column
10cm
Alkali-stabilized Protein A-derived (E. coli)
Polypropylene
3bar
2° to 8°C
HiScreen MabSelect SuRe LX Prepacked Column
1 x 4.7mL
0.3 in.
0.77cm
3.93 in.
Designed for method optimization and parameter screening for capture of monoclonal antibodies
Epoxy Activation
85μm
3 to 12
Column
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiScreen MabSelect SuRe LX

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers