Learn More
Cytiva HisPrep Column
Optimize purification of histidine-tagged recombinant proteins. Cytiva Life Sciences™ HisPrep Column is a ready to use column offering convenient scale-up purification of histidine-tagged proteins.
Brand: Cytiva 28-9365-51
Code : GE
Additional Details : Weight : 0.09990kg
Description
- Optimized for convenient scale-up purification of histidine-tagged proteins
- Compatible with a wide range of reducing agents, detergents, denaturants, and other additives
- Negligible leakage of Ni2+
- Bed Volume 20ml
- High binding capacity
- Convenient and time-saving prepacked HiPrep format with precharged Ni Sepharose 6 Fast Flow medium
- Media:
- BioProcess media
- 90μm average particle size
- Metrix 6% cross-linked agarose
- pH stability working range 3-121
- Metal ion capacity ∽15μmol Ni2+™/ml medium
- Three years shelf life
- Stable in commonly used aqueous buffers - 0.1 M HCl, 0.1 M NaOH, 8 M urea3
- Column:
- Inside Diameter is 16mm
- Material of Column hardware and tube is Polypropylene and bed support is Polymethyl-Methacrylate
- Bed support Filter
- Bed Dimensions 16 x 100mm
- Flow rate <10ml/min1
- Flow Velocity <300cm/h
- Pressure Max. 1.5 bar
Specifications
0.6 in. | |
4°C to 30°C | |
20 mL |
<10 mL/min. | |
1.6 cm | |
Each |
Safety and Handling
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : Nickel