Learn More
Thermo Scientific™ HisProbe™-HRP Conjugate
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ HisProbe-HRP is an nickel (Ni2+)-activated derivative of horseradish peroxidase (HRP). This product has been optimized for direct detection of recombinant histidine-tagged proteins and other histidine-rich proteins.
Highlights:
Fast – His-tag detection is achieved with a one-step probe incubation that eliminates the need to run a lengthy two-step primary/secondary antibody reaction protocol
Economical – no need to purchase expensive primary/secondary antibodies; considerably less costly than anti-6xHis-tag antibodies
High specificity – HisProbe-HRP can recognize his-tags independent of adjacent tags
Less steric hindrance – HisProbe-HRP is smaller than antibodies and detects his tagged fusion proteins that are often undetectable when using some anti-his-tag antibodies
Specifications
Specifications
| Description | Pierce HisProbe-HRP Conjugate |
| Product Type | HRP Conjugate |
| Detection Method | Chemiluminescence |
| Form | Solid |
| Product Line | HisProbe |
| Quantity | 2 mg |
| Reactivity | All species |
| Content And Storage | Storage: Upon receipt store product at 4°C. Product is shipped at ambient temperature. |
Safety and Handling
- HisProbe-HRP Conjugate
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Carcinogenicity Category 1A
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H360D-May damage the unborn child.
- H372-Causes damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- MIXTURE LIST-Contains : Nickel(II) chloridehexahydrate
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.