missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIST1H2AC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15953-1-AP-20UL
This item is not returnable.
View return policy
Specifications
HIST1H2AC | |
Polyclonal | |
Unconjugated | |
HIST1H2AC | |
H2A/l, H2AFL, HIST1H2AC, histone cluster 1, H2ac, Histone H2A type 1 C, Histone H2A/l | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
306962, 319164, 8334 | |
-20°C | |
Liquid |
Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C0HKE2, Q93077 | |
Hist1h2ac | |
HIST1H2AC Fusion Protein Ag8663 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |