Learn More
Histamine dihydrochloride, 99%, Thermo Scientific Chemicals
£47.34 - £2295.42
Chemical Identifiers
CAS | 56-92-8 |
---|---|
Molecular Formula | C5H11Cl2N3 |
Molecular Weight (g/mol) | 184.06 |
MDL Number | MFCD00012703 |
InChI Key | PPZMYIBUHIPZOS-UHFFFAOYSA-N |
Synonym | histamine dihydrochloride, histamine 2hcl, 2-1h-imidazol-4-yl ethanamine dihydrochloride, 1h-imidazole-4-ethanamine dihydrochloride, 1h-imidazole-4-ethanamine, dihydrochloride, peremin, ceplene, 2-1h-imidazol-4-yl ethylamine dihydrochloride, 2-4-imidazolyl ethylamine dihydrochloride, histaminedium dichloride |
PubChem CID | 5818 |
IUPAC Name | 2-(1H-imidazol-5-yl)ethanamine;dihydrochloride |
SMILES | [H+].[H+].[Cl-].[Cl-].NCCC1=CN=CN1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10583311
|
Thermo Scientific Acros
150620050 |
5 g | Glass bottle |
£47.34
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10593501
|
Thermo Scientific Acros
150620250 |
25 g | Glass bottle |
£155.78
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10503511
|
Thermo Scientific Acros
150621000 |
100 g | Glass bottle |
£541.70
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11996251
|
Thermo Scientific Acros
150625000 |
500 g | Glass bottle |
£2295.42
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
56-92-8 | |
184.06 | |
PPZMYIBUHIPZOS-UHFFFAOYSA-N | |
5818 | |
[H+].[H+].[Cl-].[Cl-].NCCC1=CN=CN1 |
C5H11Cl2N3 | |
MFCD00012703 | |
histamine dihydrochloride, histamine 2hcl, 2-1h-imidazol-4-yl ethanamine dihydrochloride, 1h-imidazole-4-ethanamine dihydrochloride, 1h-imidazole-4-ethanamine, dihydrochloride, peremin, ceplene, 2-1h-imidazol-4-yl ethylamine dihydrochloride, 2-4-imidazolyl ethylamine dihydrochloride, histaminedium dichloride | |
2-(1H-imidazol-5-yl)ethanamine;dihydrochloride |
Specifications
56-92-8 | |
White | |
Authentic | |
C5H11Cl2N3 | |
25315 | |
histamine dihydrochloride, histamine 2hcl, 2-1h-imidazol-4-yl ethanamine dihydrochloride, 1h-imidazole-4-ethanamine dihydrochloride, 1h-imidazole-4-ethanamine, dihydrochloride, peremin, ceplene, 2-1h-imidazol-4-yl ethylamine dihydrochloride, 2-4-imidazolyl ethylamine dihydrochloride, histaminedium dichloride | |
PPZMYIBUHIPZOS-UHFFFAOYSA-N | |
2-(1H-imidazol-5-yl)ethanamine;dihydrochloride | |
5818 | |
99% | |
Histamine dihydrochloride |
249.0°C to 252.0°C | |
0.5% (105°C) max. | |
0.99 | |
MFCD00012703 | |
154756 | |
Solubility in water: freely soluble in water. Other solubilities: soluble in ethanol,freely soluble in methanol | |
[H+].[H+].[Cl-].[Cl-].NCCC1=CN=CN1 | |
184.06 | |
184.07 | |
Crystalline Powder |
Safety and Handling
Causes serious eye irritation, May cause respiratory irritation, May cause allergy or asthma symptoms or breathing difficulties if inhaled, May cause an allergic skin reaction, Causes skin irritation
Serious eye damage/eye irritation (category 2), Specific target organ toxicity after single exposure (category 3), Respiratory sensitisation (category 1), Skin sensitisation (category 1), Skin corrosion/irritation (category 2)
missing translation for 'einecsNumber' : 200-298-4
- Histamine dihydrochloride
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only