Learn More
Histone Macro-H2A.1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5121059
294.00 GBP valid until 2025-03-16
Use promo code "24070" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Positive test controls include: A-549, HeLa, Rat brain. The target is usually found in the following locations: Chromosome, Nucleus. Immunogen sequence: KLEAIITPPP AKKAKSPSQK KPVSKKAGGK KGARKSKKKQ GEVSKAASAD STTEGTPADG FTVLSTKSLF LGQKLNLIHS EISNLAGFEV EAIINPTNAD IDLKDDLGNT LEKKGGKEFV EAVLELRKKN GPLEVAGAAV SAGHGLPAKF VIHCNSPVWG ADKCEELLEK TVKNCLALAD DKKLKSIAFP SIGSGRNGFP KQTAAQLILK AISSYFVSTM SSSIKTVYFV LFDSESIGIY VQEMAKLDAN
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.
Specifications
Histone Macro-H2A.1 | |
Polyclonal | |
Unconjugated | |
MACROH2A1 | |
Core histone macro-H2A.1; H2A histone family member Y; H2A histone family, member Y; H2A.y; H2A/y; H2AF12M; H2AFY; Histone H2A.y; histone macroH2A1; histone macroH2A1.1; histone macroH2A1.2; macroH2A1; MACROH2A1.1; macroH2A1.2; medulloblastoma antigen MU-MB-50.205; mH2A1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
26914, 29384, 9555 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.65 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75367, Q02874, Q9QZQ8 | |
MACROH2A1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 123-372 of human macroH2A1 (NP_6132582) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.