missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ AVB Sepharose™ Columns

Enables purification of adeno associated viruses (AAV) in one step, with high purity and excellent yields. Cytiva HiTrap™ AVB Sepharose™ Columns are pre-packed with affinity chromatography medium (resin) for simple operation with a syringe, pump or chromatographic system.

£906.00

Specifications

For Use With (Application) One Step Purification of Adeno Associated Viruses
Stability Stable to all commonly used aqueous buffers
Storage Requirements Store prepacked column at 4°C to 30°C in 20% Ethanol and Media at 4°C to 8°C in 20% Ethanol.
Binding Capacity >10▿12 genecopies/mL Chromatography Mediμm
Product Type HiTrap AVB Sepharose HP Column
View More Specs

Products 1
Product Code Brand Quantity Diameter (Metric) Inner Price Quantity & Availability  
Product Code Brand Quantity Diameter (Metric) Inner Price Quantity & Availability  
11595784
View Documents
Cytiva
28-4112-11
5 x 1 mL 7 mm
£906.00
Pack of 5
In Stock
to see stock.
Add to basket
 
Description

Description

AVB Sepharose High Performance is an affinity resin designed for the purification of adeno associated virus (AAV). It enables purification of adeno associated viruses in one step with high purity and excellent yields.
  • One step purification of adeno associated viruses
  • Affinity resin with high selectivity and low background binding
  • Large surface area to give enhanced capacity
  • Prepacked HiTrap columns for simple operation with a syringe, pump, or chromatographic system
AVB Sepharose High Performance has proven affinity for adeno associated viruses from subclasses 1, 2, 3, and 5.
  • It is expected to have affinity for most of the other adeno associated virus subclasses
  • The ligand is a 14 kD fragment from a single chain antibody, expressed in yeast
Specifications

Specifications

One Step Purification of Adeno Associated Viruses
Store prepacked column at 4°C to 30°C in 20% Ethanol and Media at 4°C to 8°C in 20% Ethanol.
HiTrap AVB Sepharose HP Column
24 to 44 μm
Polypropylene
Cross-linked agarose
14 kD recombinant protein
Stable to all commonly used aqueous buffers
>10▿12 genecopies/mL Chromatography Mediμm
<150 cm/hr
25 mm
70 psi (5 bar)
3 to 10
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiTrap AVB Sepharose High Performance 1 ml,5 x 1 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501-Dispose of contents/container to .

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Product Certifications