Learn More
Cytiva HiTrap Capto Lentil Lectin
Capto Lentil Lectin is an affinity chromatography resin for purification of glycoproteins and other molecules containing carbohydrates such as α-D-mannose and α-D-glucose or sterically related residues.
Brand:  Cytiva 17548911
Product Code. 15529244
  • £1106.00 / Pack of 5
Explore more special offers
Description
Description
  • Capto Lentil Lectin resin is based on a rigid high-flow agarose base matrix that enables high flow rates.
  • The product is available in bulk as well as in various high-throughput process development formats.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5 x 1 mL
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3