missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap Capto Phenyl ImpRes

HiTrap Capto Phenyl ImpRes is prepacked with BioProcess Capto Phenyl ImpRes resins for screening and small-scale protein purifications using hydrophobic interaction chromatography.

Brand:  Cytiva 17548411


Product Code. 15407840

  • £180.00 / Pack of 5
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket

Alert:

The temperature difference between the fluid running through the column and the ambient temperature in the room should never be greater than 20°C

This item is not returnable. View return policy

Description

Description

  • High-throughput resin for intermediate and polishing protein purification, excellent choice for process-scale purification
  • Columns for screening of selectivity, binding and elution conditions, as well as small scale purifications
  • The combined properties of high binding capacity, high flow rate, and low back pressure reduces process cycle times and improves productivity at process scale
  • Excellent chemical stability allows cleaning with all standard CIP solutions, including 1 M sodium hydroxide
  • TRUSTED_SUSTAINABILITY
    Specifications

    Specifications

    1 mL
    25 mm
    5 bar [0.5 MPa] (70 psi)
    pH 3 to 13
    Up to 220 cm/h in a 1 m diameter column with a bed height of 20 cm at 20°C; measured using process buffers with the same viscosity as water at 300 kPa
    Phenyl
    40 μm
    20% ethanol at 4°C to 30°C
    Product Suggestions

    Product Suggestions

    Videos
    Safety and Handling

    Safety and Handling

    Product Identifier
    • HiTrap Capto Phenyl ImpRes, 1 ml, 5 x 1 ml

    Signal Word
    • Warning

    Hazard Category
    • Flammable liquid Category 3

    Hazard Statement
    • H226-Flammable liquid and vapour.

    Precautionary Statement
    • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
    • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
    • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
    • P280-Wear protective gloves/protective clothing/eye protection/face protection.
    • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

    Supplemental information
    • MIXTURE LIST-Contains : ethanol
    • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.

    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers