Learn More
Cytiva HiTrap™ Capto™ SP ImpRes IEX Columns
High-throughput medium for intermediate and polishing protein purification
Brand:  Cytiva 17-5468-51
 View more versions of this product
Product Code. 11623074
  • £144.00 / Pack of 5
Explore more special offers
Description
Description
  • Combined properties of high binding capacity, high flow rate and low back pressure reduces process cycle times and improves productivity at process scale
  • Rigid agarose matrix enables high bed heights and purification of viscous samples at high flow rates
  • Screening of selectivity, binding and elution conditions, as well as small scale purifications
  • Strong sulfonate anion exchanger
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HiTrap Capto SP ImpRes, 5 x 1 mL
HiTrap Capto SP ImpRes chromatography column is packed with a strong cation exchange modern resin for high resolution, small-scale protein purification.
7 mm
5 x 1 mL
<4 mL/min.
4°C to 30°C, 0.2M Sodium Acetate in 20% Ethanol
1 mL
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva HiTrap™ Capto™ SP ImpRes IEX Columns > HiTrap Capto SP ImpRes prepacked column; Pack size: 5 x 1mL